Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGGAAGGCAAGGGAGCGCTAG[A/G]AGGTCTGCTGCTTCTGAAGCCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603243 MIM: 139311 | ||||||||||||||||||||
Literature Links: |
AMFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMFR - autocrine motility factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144.5 | 1758 | Missense Mutation | TCC,TTC | S,F 643 | NP_001135.3 | |
NM_001323511.1 | 1758 | Missense Mutation | TCC,TTC | S,F 548 | NP_001310440.1 | |
NM_001323512.1 | 1758 | Missense Mutation | TCC,TTC | S,F 675 | NP_001310441.1 | |
XM_005255890.3 | 1758 | Missense Mutation | TCC,TTC | S,F 548 | XP_005255947.1 |
GNAO1 - G protein subunit alpha o1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |