Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTGCCCCAGCTGAGCGGAGCCG[C/G]CGGGGGAGGGGGAGAGGCGGGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603213 MIM: 600999 MIM: 612033 MIM: 614386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF22 - kinesin family member 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAZ - MYC associated zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042539.2 | 714 | Missense Mutation | GCC,GGC | A,G 245 | NP_001036004.1 | |
NM_001276275.1 | 714 | Missense Mutation | GCC,GGC | A,G 222 | NP_001263204.1 | |
NM_001276276.1 | 714 | Intron | NP_001263205.1 | |||
NM_002383.3 | 714 | Missense Mutation | GCC,GGC | A,G 245 | NP_002374.2 | |
XM_006721047.3 | 714 | Missense Mutation | GCC,GGC | A,G 222 | XP_006721110.1 | |
XM_006721048.3 | 714 | Missense Mutation | GCC,GGC | A,G 245 | XP_006721111.1 |
PAGR1 - PAXIP1 associated glutamate rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRT2 - proline rich transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |