Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCTGGACCCAGAGACAGTGGAT[G/T]ATGCCTTTGCTCGTGCCTTCCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMP2 - matrix metallopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127891.2 | 336 | Missense Mutation | GAT,TAT | D,Y 92 | NP_001121363.1 | |
NM_001302508.1 | 336 | Missense Mutation | GAT,TAT | D,Y 66 | NP_001289437.1 | |
NM_001302509.1 | 336 | Missense Mutation | GAT,TAT | D,Y 66 | NP_001289438.1 | |
NM_001302510.1 | 336 | Missense Mutation | GAT,TAT | D,Y 66 | NP_001289439.1 | |
NM_004530.5 | 336 | Missense Mutation | GAT,TAT | D,Y 142 | NP_004521.1 |