Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTTGATGGACAGACCAATAAG[C/G]GTGGCCATGCTGCAGTGCGGAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605278 MIM: 614778 MIM: 179503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CES2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CES2 - carboxylesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003869.5 | 350 | Intron | NP_003860.2 | |||
NM_198061.2 | 350 | Intron | NP_932327.1 | |||
XM_011523421.2 | 350 | Intron | XP_011521723.1 | |||
XM_017023818.1 | 350 | Intron | XP_016879307.1 |
FAM96B - family with sequence similarity 96 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016062.3 | 350 | Silent Mutation | ACC,ACG | T,T 97 | NP_057146.1 |
RRAD - RRAD, Ras related glycolysis inhibitor and calcium channel regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |