Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCGCCCAGCCCTTCCGGCCGC[A/C]GCAGCACCGCGGGGAGTAGGCCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600924 MIM: 611256 MIM: 605915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFER - growth factor, augmenter of liver regeneration | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOXO1 - NADPH oxidase organizer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267721.1 | 1239 | Missense Mutation | NP_001254650.1 | |||
NM_144603.3 | 1239 | Missense Mutation | NP_653204.1 | |||
NM_172167.2 | 1239 | Missense Mutation | NP_751907.1 | |||
NM_172168.2 | 1239 | Missense Mutation | NP_751908.1 | |||
XM_005255099.4 | 1239 | Missense Mutation | XP_005255156.1 | |||
XM_017022927.1 | 1239 | Missense Mutation | XP_016878416.1 | |||
XM_017022928.1 | 1239 | Missense Mutation | XP_016878417.1 |
TBL3 - transducin beta like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006453.2 | 1239 | Intron | NP_006444.2 |