Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602349 MIM: 613577 | ||||||||||||||||||||
Literature Links: |
C16orf59 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C16orf59 - chromosome 16 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC729652 - uncharacterized LOC729652 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6768 - microRNA 6768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTN3 - netrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006181.2 | 971 | Silent Mutation | AAA,AAG | K,K 256 | NP_006172.1 |
TBC1D24 - TBC1 domain family member 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |