Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGCCACGTGACAGATGGCACA[C/T]TGGAACGTGAAGCCTTTGCAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR59 - WD repeat domain 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324171.1 | 2613 | Intron | NP_001311100.1 | |||
NM_001324172.1 | 2613 | Intron | NP_001311101.1 | |||
NM_030581.3 | 2613 | Silent Mutation | NP_085058.3 | |||
XM_005256146.3 | 2613 | Missense Mutation | XP_005256203.1 | |||
XM_011523331.2 | 2613 | Missense Mutation | XP_011521633.1 | |||
XM_011523332.2 | 2613 | Missense Mutation | XP_011521634.1 | |||
XM_017023667.1 | 2613 | Missense Mutation | XP_016879156.1 | |||
XM_017023668.1 | 2613 | Missense Mutation | XP_016879157.1 | |||
XM_017023669.1 | 2613 | Missense Mutation | XP_016879158.1 | |||
XM_017023670.1 | 2613 | Missense Mutation | XP_016879159.1 |