Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTCGAAGGCCAGAAGGGTCAG[C/T]GAGAGCAGCCTGTCTGCTAGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA2L - spermatogenesis associated 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS9D1 - VPS9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004913.2 | 3270 | Silent Mutation | TCA,TCG | S,S 424 | NP_004904.2 | |
XM_005256329.4 | 3270 | Silent Mutation | TCA,TCG | S,S 425 | XP_005256386.1 | |
XM_005256330.4 | 3270 | Silent Mutation | TCA,TCG | S,S 355 | XP_005256387.1 | |
XM_005256331.2 | 3270 | Silent Mutation | TCA,TCG | S,S 259 | XP_005256388.1 | |
XM_006721350.2 | 3270 | Silent Mutation | TCA,TCG | S,S 425 | XP_006721413.1 | |
XM_011523476.2 | 3270 | Silent Mutation | TCA,TCG | S,S 424 | XP_011521778.1 | |
XM_011523477.2 | 3270 | Silent Mutation | TCA,TCG | S,S 391 | XP_011521779.1 | |
XM_011523478.2 | 3270 | Silent Mutation | TCA,TCG | S,S 390 | XP_011521780.1 | |
XM_011523479.2 | 3270 | Silent Mutation | TCA,TCG | S,S 425 | XP_011521781.1 | |
XM_011523480.2 | 3270 | Silent Mutation | TCA,TCG | S,S 355 | XP_011521782.1 |
VPS9D1-AS1 - VPS9D1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |