Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACCTTGTACTTGTCGTAGATG[C/G]CTGCCCGCCGCTCGGGGTCCTCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604853 | ||||||||||||||||||||
Literature Links: |
MRPL28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL28 - mitochondrial ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006428.4 | 590 | Missense Mutation | GCC,GGC | A,G 186 | NP_006419.2 | |
XM_005255041.1 | 590 | Missense Mutation | GCC,GGC | A,G 186 | XP_005255098.1 | |
XM_011522351.2 | 590 | Missense Mutation | GCC,GGC | A,G 186 | XP_011520653.1 |
TMEM8A - transmembrane protein 8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |