Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAAGGCGGCTTCGGGCTGCGAA[C/G]GGCTTCAGGGACCACCCACCCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615624 MIM: 606195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRNDE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRNDE - colorectal neoplasia differentially expressed (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRX5 - iroquois homeobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252197.1 | 1134 | Silent Mutation | CGG,GGG | R,G 230 | NP_001239126.1 | |
NM_005853.5 | 1134 | Silent Mutation | CGG,GGG | R,G 231 | NP_005844.4 | |
XM_011522809.1 | 1134 | Silent Mutation | CGG,GGG | R,G 161 | XP_011521111.1 |