Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGCGTCCACGGCGTTGGTCAGT[A/G]TGGTCAGGTTGGCCTCACACTGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602941 MIM: 118890 | ||||||||||||||||||||
Literature Links: |
BCAR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAR1 - BCAR1, Cas family scaffolding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170714.1 | 2844 | Missense Mutation | ACA,ATA | T,I 803 | NP_001164185.1 | |
NM_001170715.1 | 2844 | Missense Mutation | ACA,ATA | T,I 775 | NP_001164186.1 | |
NM_001170716.1 | 2844 | Missense Mutation | ACA,ATA | T,I 775 | NP_001164187.1 | |
NM_001170717.1 | 2844 | Missense Mutation | ACA,ATA | T,I 775 | NP_001164188.1 | |
NM_001170718.1 | 2844 | Missense Mutation | ACA,ATA | T,I 757 | NP_001164189.1 | |
NM_001170719.1 | 2844 | Missense Mutation | ACA,ATA | T,I 755 | NP_001164190.1 | |
NM_001170720.1 | 2844 | Missense Mutation | ACA,ATA | T,I 609 | NP_001164191.1 | |
NM_001170721.1 | 2844 | Missense Mutation | ACA,ATA | T,I 547 | NP_001164192.1 | |
NM_014567.3 | 2844 | Missense Mutation | ACA,ATA | T,I 757 | NP_055382.2 | |
XM_005256258.4 | 2844 | Missense Mutation | ACA,ATA | T,I 783 | XP_005256315.1 | |
XM_005256259.4 | 2844 | Missense Mutation | ACA,ATA | T,I 757 | XP_005256316.1 | |
XM_005256260.4 | 2844 | Missense Mutation | ACA,ATA | T,I 728 | XP_005256317.1 | |
XM_011523475.2 | 2844 | Missense Mutation | ACA,ATA | T,I 728 | XP_011521777.1 | |
XM_017023897.1 | 2844 | Missense Mutation | ACA,ATA | T,I 803 | XP_016879386.1 | |
XM_017023898.1 | 2844 | Missense Mutation | ACA,ATA | T,I 773 | XP_016879387.1 | |
XM_017023899.1 | 2844 | Missense Mutation | ACA,ATA | T,I 710 | XP_016879388.1 |
CTRB1 - chymotrypsinogen B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100506281 - uncharacterized LOC100506281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |