Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGCGCTTCAGCCTCATCACAG[A/C]CGGGTGGTCCTCTCCACGGCGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603365 MIM: 613199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIRIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIRIP3 - HIRA interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197323.1 | 1764 | Missense Mutation | GGC,GTC | G,V 112 | NP_001184252.1 | |
NM_003609.4 | 1764 | Missense Mutation | GCT,TCT | A,S 425 | NP_003600.2 |
INO80E - INO80 complex subunit E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAOK2 - TAO kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |