Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAACCTGTGAGTCCTGAGGCATT[C/T]AGATCCTCCTTGGTCTGCACCACGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607490 MIM: 612060 | ||||||||||||||||||||
Literature Links: |
LDHD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDHD - lactate dehydrogenase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153486.3 | 1210 | Silent Mutation | CTA,CTG | L,L 409 | NP_705690.2 | |
NM_194436.2 | 1210 | Silent Mutation | CTA,CTG | L,L 386 | NP_919417.1 |
ZNRF1 - zinc and ring finger 1, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |