Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600659 MIM: 606422 MIM: 614117 MIM: 613701 | ||||||||||||||||||||
Literature Links: |
E2F4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
E2F4 - E2F transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001950.3 | 353 | Intron | NP_001941.2 |
ELMO3 - engulfment and cell motility 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024712.3 | 353 | Silent Mutation | CTG,GTG | L,V 104 | NP_078988.2 |
EXOC3L1 - exocyst complex component 3 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC29 - leucine rich repeat containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR328 - microRNA 328 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |