Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGCAGGGGCAGCAGGTGCACAG[A/G]CTGTGGCTGGCGGCCCTGCCACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118888 MIM: 606967 MIM: 176847 MIM: 604119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTRL - chymotrypsin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCAT - lecithin-cholesterol acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000229.1 | 1179 | Missense Mutation | CCT,TCT | P,S 390 | NP_000220.1 |
PSMB10 - proteasome subunit beta 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A4 - solute carrier family 12 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |