Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCAGGGACAGTGGCCACCATG[A/C]GTAAGCCCCGCTGCTCCCTGCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMP25 - matrix metallopeptidase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022468.4 | 230 | Missense Mutation | AGT,CGT | S,R 86 | NP_071913.1 | |
XM_011522602.2 | 230 | Missense Mutation | AGT,CGT | S,R 52 | XP_011520904.1 | |
XM_011522604.2 | 230 | Missense Mutation | AGT,CGT | S,R 2 | XP_011520906.1 | |
XM_011522605.2 | 230 | Intron | XP_011520907.1 | |||
XM_017023561.1 | 230 | Missense Mutation | AGT,CGT | S,R 86 | XP_016879050.1 |
MMP25-AS1 - MMP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |