Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609377 MIM: 610859 MIM: 607484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACD - adrenocortical dysplasia homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf86 - chromosome 16 open reading frame 86 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CARMIL2 - capping protein regulator and myosin 1 linker 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ENKD1 - enkurin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARD6A - par-6 family cell polarity regulator alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037281.1 | 546 | Missense Mutation | CAT,TAT | H,Y 162 | NP_001032358.1 | |
NM_016948.2 | 546 | Missense Mutation | CAT,TAT | H,Y 163 | NP_058644.1 | |
XM_005255977.3 | 546 | Missense Mutation | CAT,TAT | H,Y 174 | XP_005256034.1 | |
XM_011523095.2 | 546 | Missense Mutation | CAT,TAT | H,Y 173 | XP_011521397.1 | |
XM_011523096.2 | 546 | Missense Mutation | CAT,TAT | H,Y 170 | XP_011521398.1 | |
XM_017023261.1 | 546 | Missense Mutation | CAT,TAT | H,Y 158 | XP_016878750.1 |