Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACCTGGCCGGTTGAAGTCCATG[C/G]ACGGCTGTTTGCAGTCCTGGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609604 | ||||||||||||||||||||
Literature Links: |
MAP1LC3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1LC3B - microtubule associated protein 1 light chain 3 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC14 - zinc finger CCHC-type containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015144.2 | 2992 | Missense Mutation | TCC,TGC | S,C 925 | NP_055959.1 | |
XM_005255858.3 | 2992 | Missense Mutation | TCC,TGC | S,C 925 | XP_005255915.2 | |
XM_011522964.2 | 2992 | Intron | XP_011521266.1 | |||
XM_017023082.1 | 2992 | Missense Mutation | TCC,TGC | S,C 889 | XP_016878571.1 |