Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGACTTCAAACTCGTCTTTGGC[A/G]AGGACGGGGCGCCGGCGCCGCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609707 MIM: 602698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUS2 - dihydrouridine synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFATC3 - nuclear factor of activated T-cells 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004555.3 | 374 | Missense Mutation | AAG,GAG | K,E 20 | NP_004546.1 | |
NM_173163.2 | 374 | Missense Mutation | AAG,GAG | K,E 20 | NP_775186.1 | |
NM_173165.2 | 374 | Missense Mutation | AAG,GAG | K,E 20 | NP_775188.1 |