Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTGTACAGGTCTTAAAAGAGA[A/C]GCCATATCCCATTGCTGAAGTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC11 - ATP binding cassette subfamily C member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LONP2 - lon peptidase 2, peroxisomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300948.1 | 572 | Missense Mutation | AAG,ACG | K,T 117 | NP_001287877.1 | |
NM_031490.3 | 572 | Missense Mutation | AAG,ACG | K,T 117 | NP_113678.2 | |
XM_017023755.1 | 572 | Missense Mutation | AAG,ACG | K,T 117 | XP_016879244.1 | |
XM_017023756.1 | 572 | Missense Mutation | AAG,ACG | K,T 117 | XP_016879245.1 |