Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTTTCTGTTATCCTACAAAGC[A/G]AAGAATAAGGAGGAGGCCCCGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600227 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA17P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA17P - ATP binding cassette subfamily A member 17, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCNF - cyclin F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323538.1 | 310 | UTR 5 | NP_001310467.1 | |||
NM_001761.2 | 310 | Missense Mutation | CAA,CGA | Q,R 21 | NP_001752.2 | |
XM_017023822.1 | 310 | Missense Mutation | CAA,CGA | Q,R 102 | XP_016879311.1 | |
XM_017023823.1 | 310 | Missense Mutation | CAA,CGA | Q,R 102 | XP_016879312.1 |