Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGACCAGAATGGAAACGACCTTC[A/G]GACCCGCCTTTTCAGCCGTCACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606692 | ||||||||||||||||||||
Literature Links: |
TRAF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAF7 - TNF receptor associated factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032271.2 | 244 | Missense Mutation | AGA,GGA | R,G 35 | NP_115647.2 | |
XM_005255627.4 | 244 | Missense Mutation | AGA,GGA | R,G 35 | XP_005255684.1 | |
XM_011522700.1 | 244 | Missense Mutation | AGA,GGA | R,G 6 | XP_011521002.1 |