Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGACCCTCCACCGGGCTCGCC[A/G]GTCCGTCGGACTTTTGGCTGAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104155 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFHX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFHX3 - zinc finger homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164766.1 | 8411 | Missense Mutation | NP_001158238.1 | |||
NM_006885.3 | 8411 | Missense Mutation | NP_008816.3 | |||
XM_005255957.3 | 8411 | Missense Mutation | XP_005256014.1 | |||
XM_017023251.1 | 8411 | Missense Mutation | XP_016878740.1 |