Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGGGGCCTCCCTTATCTCTCT[A/G]CCTCCATGGCAGGGTGGGCCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf96 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf96 - chromosome 16 open reading frame 96 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGRN1 - mahogunin ring finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBALD1 - UBA like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145253.2 | 653 | Missense Mutation | GCA,GTA | A,V 175 | NP_660296.1 | |
XM_017022937.1 | 653 | Missense Mutation | GCA,GTA | A,V 150 | XP_016878426.1 |