Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTGCGCGCTGTGCTCCTGCA[A/C]CAAGGCCACGTCTAGGAAGCGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610264 MIM: 613276 | ||||||||||||||||||||
Literature Links: |
TEPP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEPP - testis, prostate and placenta expressed | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USB1 - U6 snRNA biogenesis phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF319 - zinc finger protein 319 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020807.2 | 2292 | Missense Mutation | GTG,TTG | V,L 557 | NP_065858.1 | |
XM_005256069.3 | 2292 | Missense Mutation | GTG,TTG | V,L 557 | XP_005256126.1 |