Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGCTGGGCCCTGCGCCTTCCC[C/G]TAGGCCAATTCTATAATCAGGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607700 | ||||||||||||||||||||
Literature Links: |
RNF40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF40 - ring finger protein 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207033.1 | 2306 | Intron | NP_001193962.1 | |||
NM_001207034.1 | 2306 | Intron | NP_001193963.1 | |||
NM_001286572.2 | 2306 | Intron | NP_001273501.1 | |||
NM_014771.3 | 2306 | Intron | NP_055586.1 | |||
XM_011545997.1 | 2306 | Intron | XP_011544299.1 |
ZNF629 - zinc finger protein 629 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080417.1 | 2306 | Missense Mutation | CGG,GGG | R,G 700 | NP_001073886.1 | |
XM_005255222.3 | 2306 | Missense Mutation | CGG,GGG | R,G 686 | XP_005255279.1 |