Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGGAACCTGAAGCTCTTTGAA[A/G]GGTATCTCTGCACCCTGAGAATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600227 | ||||||||||||||||||||
Literature Links: |
ABCA17P PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA17P - ATP binding cassette subfamily A member 17, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCNF - cyclin F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323538.1 | 525 | Intron | NP_001310467.1 | |||
NM_001761.2 | 525 | Missense Mutation | AGG,GGG | R,G 93 | NP_001752.2 | |
XM_017023822.1 | 525 | Missense Mutation | AGG,GGG | R,G 174 | XP_016879311.1 | |
XM_017023823.1 | 525 | Missense Mutation | AGG,GGG | R,G 174 | XP_016879312.1 |