Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCATGTGAAGGGCGCCAATGCCG[G/T]GGTCCAAACCTTCCTGGGAATTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605278 MIM: 614778 | ||||||||||||||||||||
Literature Links: |
CES2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CES2 - carboxylesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003869.5 | 1353 | Missense Mutation | GGG,GTG | G,V 118 | NP_003860.2 | |
NM_198061.2 | 1353 | Missense Mutation | GGG,GTG | G,V 118 | NP_932327.1 | |
XM_011523421.2 | 1353 | UTR 5 | XP_011521723.1 | |||
XM_017023818.1 | 1353 | UTR 5 | XP_016879307.1 |
FAM96B - family with sequence similarity 96 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |