Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGATGACCGTCAGGGCGTCCTC[A/G]GAGGCCTGCCGCTTCGCCTCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBFA2T3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBFA2T3 - CBFA2/RUNX1 translocation partner 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005187.5 | 1572 | Silent Mutation | TCC,TCT | S,S 539 | NP_005178.4 | |
NM_175931.2 | 1572 | Silent Mutation | TCC,TCT | S,S 453 | NP_787127.1 | |
XM_005256323.4 | 1572 | Silent Mutation | TCC,TCT | S,S 514 | XP_005256380.1 | |
XM_011523419.2 | 1572 | Silent Mutation | TCC,TCT | S,S 539 | XP_011521721.1 | |
XM_017023811.1 | 1572 | Intron | XP_016879300.1 |
LOC101927793 - uncharacterized LOC101927793 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPN1L - poly(A) binding protein nuclear 1 like (cytoplasmic) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |