Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCCTTCTGTGGCTTGACTTGCA[A/G]GTTCAGGTTTGGCTCCTCCCTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616662 | ||||||||||||||||||||
Literature Links: |
THUMPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THUMPD1 - THUMP domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304550.1 | 1177 | Intron | NP_001291479.1 | |||
NM_017736.4 | 1177 | Intron | NP_060206.2 | |||
XM_011545891.1 | 1177 | Intron | XP_011544193.1 | |||
XM_017023433.1 | 1177 | Missense Mutation | CCT,CTT | P,L 337 | XP_016878922.1 |