Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCTCTCTCTAGACTCCAGTTGC[C/T]TCTCTTCTCCACCTGTGAACTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMC7 - transmembrane channel like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160364.2 | 322 | UTR 5 | NP_001153836.1 | |||
NM_001300732.1 | 322 | Missense Mutation | CTC,TTC | L,F 32 | NP_001287661.1 | |
NM_001324263.1 | 322 | UTR 5 | NP_001311192.1 | |||
NM_001324265.1 | 322 | Missense Mutation | CTC,TTC | L,F 32 | NP_001311194.1 | |
NM_001324268.1 | 322 | UTR 5 | NP_001311197.1 | |||
NM_024847.4 | 322 | Missense Mutation | CTC,TTC | L,F 32 | NP_079123.3 |