Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCATGGCCCAGCCGCCGCCCGAC[A/G]TGGAGGGGGACGACTGTCTCCCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615839 MIM: 601818 | ||||||||||||||||||||
Literature Links: |
DECR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DECR2 - 2,4-dienoyl-CoA reductase 2, peroxisomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020664.3 | 128 | Missense Mutation | ATG,GTG | M,V 8 | NP_065715.1 |
LOC100134368 - uncharacterized LOC100134368 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME4 - NME/NM23 nucleoside diphosphate kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |