Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTACGCTGTGCCCAGCGTACC[A/G]CGGGCGCGCCGGACCCCGCTTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614117 MIM: 602438 MIM: 605235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC3L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC3L1 - exocyst complex component 3 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSF4 - heat shock transcription factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0895L - KIAA0895 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL3 - nucleolar protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185057.2 | 452 | Missense Mutation | ACG,GCG | T,A 84 | NP_001171986.1 | |
NM_001276307.1 | 452 | Missense Mutation | ACG,GCG | T,A 84 | NP_001263236.1 | |
NM_001276309.1 | 452 | Missense Mutation | ACG,GCG | T,A 84 | NP_001263238.1 | |
NM_001276311.1 | 452 | Intron | NP_001263240.1 | |||
NM_001276312.1 | 452 | Missense Mutation | ACG,GCG | T,A 84 | NP_001263241.1 | |
NM_001276319.1 | 452 | Missense Mutation | ACG,GCG | T,A 146 | NP_001263248.1 | |
NM_003946.6 | 452 | Missense Mutation | ACG,GCG | T,A 84 | NP_003937.1 | |
XM_005256217.2 | 452 | Missense Mutation | ACG,GCG | T,A 146 | XP_005256274.1 | |
XM_005256219.3 | 452 | Intron | XP_005256276.1 | |||
XM_011523424.2 | 452 | Missense Mutation | ACG,GCG | T,A 84 | XP_011521726.1 | |
XM_017023843.1 | 452 | Missense Mutation | ACG,GCG | T,A 146 | XP_016879332.1 |