Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGGTGGGTGCGGCTCAGCTCC[C/T]ACCACTCTTGCACGTGCTGGTACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613374 MIM: 601292 | ||||||||||||||||||||
Literature Links: |
SGF29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGF29 - SAGA complex associated factor 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT1A2 - sulfotransferase family 1A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001054.3 | 756 | Missense Mutation | NP_001045.1 | |||
NM_177528.2 | 756 | Missense Mutation | NP_803564.1 | |||
XM_006721077.3 | 756 | Intron | XP_006721140.1 | |||
XM_017023600.1 | 756 | Nonsense Mutation | XP_016879089.1 | |||
XM_017023601.1 | 756 | Nonsense Mutation | XP_016879090.1 | |||
XM_017023602.1 | 756 | Nonsense Mutation | XP_016879091.1 | |||
XM_017023603.1 | 756 | Intron | XP_016879092.1 |