Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAAGTTGATCAACCAGGTTCA[A/G]TATGTTTGTGGAAGACGCAAAGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NSMCE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSMCE1 - NSE1 homolog, SMC5-SMC6 complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145080.3 | 504 | Silent Mutation | CTG,TTG | L,L 130 | NP_659547.2 | |
XM_006721023.3 | 504 | Silent Mutation | CTG,TTG | L,L 130 | XP_006721086.1 | |
XM_011545761.2 | 504 | Silent Mutation | CTG,TTG | L,L 167 | XP_011544063.1 | |
XM_011545762.1 | 504 | Silent Mutation | CTG,TTG | L,L 34 | XP_011544064.1 | |
XM_017023040.1 | 504 | Silent Mutation | CTG,TTG | L,L 34 | XP_016878529.1 | |
XM_017023041.1 | 504 | Silent Mutation | CTG,TTG | L,L 34 | XP_016878530.1 | |
XM_017023042.1 | 504 | Silent Mutation | CTG,TTG | L,L 34 | XP_016878531.1 |