Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCCCCACCTACCCGCTCCAGA[C/T]TGAGGATGGTGGCCATCTGGGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606976 MIM: 608675 | ||||||||||||||||||||
Literature Links: |
COG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COG4 - component of oligomeric golgi complex 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195139.1 | 2396 | Missense Mutation | AAT,AGT | N,S 721 | NP_001182068.1 | |
NM_015386.2 | 2396 | Missense Mutation | AAT,AGT | N,S 742 | NP_056201.2 | |
XM_011522981.2 | 2396 | Missense Mutation | AAT,AGT | N,S 600 | XP_011521283.1 |
FUK - fucokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |