Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAACATGCGGAGGTCCTCAGGGT[A/T]CTGGGCTGCGGGTACACTGAGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 156565 MIM: 600928 | ||||||||||||||||||||
Literature Links: |
MPG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPG - N-methylpurine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPRL3 - NPR3 like, GATOR1 complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039476.2 | 1840 | Missense Mutation | AAC,TAC | N,Y 327 | NP_001034565.1 | |
NM_001077350.2 | 1840 | Missense Mutation | AAC,TAC | N,Y 506 | NP_001070818.1 | |
NM_001243247.1 | 1840 | Missense Mutation | AAC,TAC | N,Y 428 | NP_001230176.1 | |
NM_001243248.1 | 1840 | Missense Mutation | AAC,TAC | N,Y 481 | NP_001230177.1 | |
NM_001243249.1 | 1840 | Missense Mutation | AAC,TAC | N,Y 481 | NP_001230178.1 |