Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGAGCTACGACTACCTGCTCAA[A/G]TTCCTGCTGGTGGGCGACAGCGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929280 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929280 - uncharacterized LOC101929280 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGQ - phosphatidylinositol glycan anchor biosynthesis class Q | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB40C - RAB40C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172663.1 | 122 | Silent Mutation | AAA,AAG | K,K 16 | NP_001166134.1 | |
NM_001172664.1 | 122 | Silent Mutation | AAA,AAG | K,K 16 | NP_001166135.1 | |
NM_001172665.1 | 122 | Silent Mutation | AAA,AAG | K,K 16 | NP_001166136.1 | |
NM_001172666.1 | 122 | Silent Mutation | AAA,AAG | K,K 16 | NP_001166137.1 | |
NM_021168.4 | 122 | Silent Mutation | AAA,AAG | K,K 16 | NP_066991.3 |