Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCTTCTCTGTCCAGGTATACC[A/G]CTGGGCCGACCACTCAAGCACGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL36 - kelch like family member 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303451.1 | 226 | Missense Mutation | CAC,CGC | H,R 24 | NP_001290380.1 | |
NM_024731.3 | 226 | Missense Mutation | CAC,CGC | H,R 24 | NP_079007.2 | |
XM_005256149.2 | 226 | Missense Mutation | CAC,CGC | H,R 24 | XP_005256206.1 | |
XM_017023674.1 | 226 | Missense Mutation | CAC,CGC | H,R 24 | XP_016879163.1 | |
XM_017023675.1 | 226 | Missense Mutation | CAC,CGC | H,R 24 | XP_016879164.1 |