Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAGACGCAGAGGGGGCATCTC[C/T]TTCCTCCCTGGGGCTACGGTAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf71 - chromosome 16 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139170.2 | 1453 | Missense Mutation | CCT,CTT | P,L 421 | NP_631909.2 | |
XM_005255144.3 | 1453 | Missense Mutation | CCT,CTT | P,L 283 | XP_005255201.1 | |
XM_017022975.1 | 1453 | Missense Mutation | CCT,CTT | P,L 444 | XP_016878464.1 | |
XM_017022976.1 | 1453 | Missense Mutation | CCT,CTT | P,L 421 | XP_016878465.1 |
ZNF500 - zinc finger protein 500 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303450.1 | 1453 | Intron | NP_001290379.1 | |||
NM_021646.3 | 1453 | Intron | NP_067678.1 | |||
XM_005255243.3 | 1453 | Intron | XP_005255300.1 | |||
XM_011522453.2 | 1453 | Intron | XP_011520755.1 | |||
XM_011522454.2 | 1453 | Intron | XP_011520756.1 | |||
XM_017023121.1 | 1453 | Intron | XP_016878610.1 |