Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGGTCGCTGACCAGACGCTCCC[C/T]GCTGACGAACAGGAAGTGGGACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST6 - heparan sulfate-glucosamine 3-sulfotransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009606.3 | 971 | Missense Mutation | AGG,GGG | R,G 249 | NP_001009606.3 | |
XM_011522608.2 | 971 | Missense Mutation | AGG,GGG | R,G 124 | XP_011520910.1 | |
XM_011522609.2 | 971 | Missense Mutation | AGG,GGG | R,G 109 | XP_011520911.1 |