Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTCTCATATGCTTCTTCACTC[A/G]TAAGTTCATCTAGTTCTGAAGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 238330 | ||||||||||||||||||||
Literature Links: |
C16orf46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C16orf46 - chromosome 16 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCSH - glycine cleavage system protein H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004483.4 | 573 | Missense Mutation | ACG,ATG | T,M 159 | NP_004474.2 | |
XM_017023136.1 | 573 | Intron | XP_016878625.1 | |||
XM_017023137.1 | 573 | Intron | XP_016878626.1 |