Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGTCCGGTAGTCAGAGATGCT[A/G]CATCCAAACAGCAGGTTAAACTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616434 MIM: 607895 | ||||||||||||||||||||
Literature Links: |
IST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IST1 - IST1, ESCRT-III associated factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD1L3 - polycystin 1 like 3, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181536.1 | 4845 | Silent Mutation | TGC,TGT | C,C 1615 | NP_853514.1 | |
XM_017023201.1 | 4845 | Intron | XP_016878690.1 | |||
XM_017023202.1 | 4845 | Intron | XP_016878691.1 | |||
XM_017023203.1 | 4845 | Intron | XP_016878692.1 | |||
XM_017023204.1 | 4845 | Intron | XP_016878693.1 |