Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGCTCATCTAGACTGTAGTAT[C/T]GGTGAACAATTTTCTCGGCGGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPPED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPPED1 - calcineurin like phosphoesterase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099455.1 | 557 | Missense Mutation | CAA,CGA | Q,R 149 | NP_001092925.1 | |
NM_018340.2 | 557 | Missense Mutation | CAA,CGA | Q,R 291 | NP_060810.2 |