Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAAACTGAGCTGGGTGGGGTAT[A/C]AGTTACAGGGGCGAGAGCTTCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600305 MIM: 606447 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECI1 - enoyl-CoA delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNPS1 - RNA binding protein with serine rich domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286625.1 | 1425 | Intron | NP_001273554.1 | |||
NM_001286626.1 | 1425 | UTR 3 | NP_001273555.1 | |||
NM_001286627.1 | 1425 | Missense Mutation | GAT,TAT | D,Y 247 | NP_001273556.1 | |
NM_006711.4 | 1425 | Intron | NP_006702.1 | |||
NM_080594.3 | 1425 | UTR 3 | NP_542161.1 | |||
XM_005255048.1 | 1425 | Intron | XP_005255105.1 | |||
XM_005255049.3 | 1425 | Intron | XP_005255106.1 | |||
XM_017022874.1 | 1425 | Intron | XP_016878363.1 |