Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGACAGCTCATCTGCGCGGCTGCT[C/T]TCGTAGGCGCGGGAAGCACAGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600924 MIM: 611256 MIM: 605915 | ||||||||||||||||||||
Literature Links: |
GFER PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFER - growth factor, augmenter of liver regeneration | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOXO1 - NADPH oxidase organizer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267721.1 | 1114 | Silent Mutation | GAA,GAG | E,E 248 | NP_001254650.1 | |
NM_144603.3 | 1114 | Silent Mutation | GAA,GAG | E,E 243 | NP_653204.1 | |
NM_172167.2 | 1114 | Silent Mutation | GAA,GAG | E,E 244 | NP_751907.1 | |
NM_172168.2 | 1114 | Silent Mutation | GAA,GAG | E,E 249 | NP_751908.1 | |
XM_005255099.4 | 1114 | Silent Mutation | GAA,GAG | E,E 249 | XP_005255156.1 | |
XM_017022927.1 | 1114 | Silent Mutation | GAA,GAG | E,E 249 | XP_016878416.1 | |
XM_017022928.1 | 1114 | Silent Mutation | GAA,GAG | E,E 249 | XP_016878417.1 |
TBL3 - transducin beta like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006453.2 | 1114 | Intron | NP_006444.2 |