Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTCCTGCGTGGTGCAGAGCACG[A/G]GCCAGATGCAGTGCAAGGTGTACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615798 MIM: 615799 MIM: 605914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN6 - claudin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLDN9 - claudin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020982.3 | 1085 | Missense Mutation | AGC,GGC | S,G 60 | NP_066192.1 |
HCFC1R1 - host cell factor C1 regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF12A - TNF receptor superfamily member 12A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |