Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCCCCCAGGAGATAGTGGCTG[C/G]CAACCAAAGGGGCAGCCTCTACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120980 | ||||||||||||||||||||
Literature Links: |
ITGAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGAM - integrin subunit alpha M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000632.3 | 35 | Missense Mutation | GCC,GGC | A,G 57 | NP_000623.2 | |
NM_001145808.1 | 35 | Missense Mutation | GCC,GGC | A,G 57 | NP_001139280.1 | |
XM_006721045.1 | 35 | Missense Mutation | GCC,GGC | A,G 57 | XP_006721108.1 | |
XM_011545850.2 | 35 | UTR 5 | XP_011544152.1 | |||
XM_011545851.2 | 35 | Missense Mutation | GCC,GGC | A,G 57 | XP_011544153.1 | |
XM_017023216.1 | 35 | Missense Mutation | GCC,GGC | A,G 57 | XP_016878705.1 |