Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCCTCCTCACCCCCCGCCTCG[C/G]CTGCCCCTTCCAAGGGACCAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF668 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF668 - zinc finger protein 668 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172668.1 | 1816 | Missense Mutation | GCC,GGC | A,G 502 | NP_001166139.1 | |
NM_001172669.1 | 1816 | Missense Mutation | GCC,GGC | A,G 525 | NP_001166140.1 | |
NM_001172670.1 | 1816 | Missense Mutation | GCC,GGC | A,G 502 | NP_001166141.1 | |
NM_024706.4 | 1816 | Missense Mutation | GCC,GGC | A,G 502 | NP_078982.3 |