Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTGATTGATGGCTCTGGTAGC[A/G]TCATCCCACATGACTTTCGGCGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGAM - integrin subunit alpha M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000632.3 | 375 | Missense Mutation | ATC,GTC | I,V 161 | NP_000623.2 | |
NM_001145808.1 | 375 | Missense Mutation | ATC,GTC | I,V 161 | NP_001139280.1 | |
XM_006721045.1 | 375 | Missense Mutation | ATC,GTC | I,V 161 | XP_006721108.1 | |
XM_011545850.2 | 375 | Missense Mutation | ATC,GTC | I,V 99 | XP_011544152.1 | |
XM_011545851.2 | 375 | Missense Mutation | ATC,GTC | I,V 161 | XP_011544153.1 | |
XM_017023216.1 | 375 | Missense Mutation | ATC,GTC | I,V 161 | XP_016878705.1 |